4-Aminopiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperidin-4-amine

M-Clarity™ Qualitätsstufe = MQ100

561479-25G 561479-5G
SIAL561479-25GEA 134 EUR
SIAL561479-25G SIAL561479-5G
4-Aminopiperidin, Sigma-Aldrich®
4-Aminopiperidin
Formel: C₅H₁₂N₂
Molecular Weight: 100.16 g/mol
MDL: MFCD02179399
CAS-Nummer: 13035-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR