4-Amino-1-benzylpiperidin

Lieferant: FLUOROCHEM

Synonyme: 4-Amino-N-benzylpiperidine

008925-5G 008925-100G 008925-25G
FLUO008925-5GEA 0 EUR
FLUO008925-5G FLUO008925-100G FLUO008925-25G
4-Amino-1-benzylpiperidin
4-Amino-1-benzylpiperidin
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
MDL: MFCD00006504
CAS-Nummer: 50541-93-0
EINECS: 256-620-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR