1-Methyl-2-piperidinmethanol ≥98%

Lieferant: Thermo Scientific
Achtung

B22187.06 B22187.14 B22187.22
B22187.06EA 24.2 EUR
B22187.06 B22187.14 B22187.22
1-Methyl-2-piperidinmethanol ≥98%
1-Methyl-2-piperidinmethanol

Purity: 98% CAS No.: 20845-34-5 Molecular Formula: C7H15NO Formula Weight: 129.20 Flash Point: 81°(177°F) Refractive Index: 1.4830 Boiling Point: 79-80°/7mm Beilstein Reference: 103612 MDL No.: MFCD00006494

Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Siedepunkt: 79…80 °C (7 mmHg)
Dichte: 0,984
Flammpunkt: 81 °C (177 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006494
CAS-Nummer: 20845-34-5
EINECS: 244-078-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR