2,2,6,6-Tetramethylpiperidin-1-oxyl (TEMPO, freies Radikal)
Lieferant: CHEMPUR
Synonyme:
TEMPO
FL13714.POR
013714-100G
013714-25G
CHMPFL13714.POREA
0
EUR
CHMPFL13714.POR
FLUO013714-100G
FLUO013714-25G
2,2,6,6-Tetramethylpiperidin-1-oxyl (TEMPO, freies Radikal)
2,2,6,6-Tetramethylpiperidin-1-oxyl (TEMPO)
Formel:
C₉H₁₈NO Molecular Weight: 156,25 g/mol |
MDL:
MFCD00009599 CAS-Nummer: 2564-83-2 EINECS: 219-888-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...