4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine
M-Clarity™ Qualitätsstufe = MQ200
163945-1G
163945-5G
SIAL163945-1GEA
136
EUR
SIAL163945-1G
SIAL163945-5G
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO), Sigma-Aldrich®
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO)
Formel:
C₉H₁₉N₂O Molecular Weight: 171.26 g/mol |
MDL:
MFCD00006479 CAS-Nummer: 14691-88-4 EINECS: 238-738-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...