4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol

M-Clarity™ Qualitätsstufe = MQ200

176141-5G 176141-25G 176141-1G
SIAL176141-5GEA 105 EUR
SIAL176141-5G SIAL176141-25G SIAL176141-1G
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL), Sigma-Aldrich®
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL)
Formel: C₉H₁₈NO₂
Molecular Weight: 172,25 g/mol
MDL: MFCD00006478
CAS-Nummer: 2226-96-2
EINECS: 218-760-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR