2,2,6,6-Tetramethyl-4-piperidinol 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

151940250 151942500 151940050
ACRO151940250EA 86.7 EUR
ACRO151940250 ACRO151942500 ACRO151940050
2,2,6,6-Tetramethyl-4-piperidinol 99%
2,2,6,6-Tetramethyl-4-piperidinol
Formel: C₉H₁₉NO
Molecular Weight: 157,26 g/mol
Schmelzpunkt: 129…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005983
CAS-Nummer: 2403-88-5
EINECS: 219-291-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 129 to 133 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR