2,2,6,6-Tetramethyl-4-piperidinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

M-Clarity™ Qualitätsstufe = MQ100

115746-10G 115746-25G
SIAL115746-10GEA 74 EUR
SIAL115746-10G SIAL115746-25G
2,2,6,6-Tetramethyl-4-piperidinol, Sigma-Aldrich®
2,2,6,6-Tetramethyl-4-piperidinol
Formel: C₉H₁₉NO
Molecular Weight: 157,26 g/mol
MDL: MFCD00005983
CAS-Nummer: 2403-88-5
EINECS: 219-291-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR