Cumarin-3-carbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Oxo-2H-chromene-3-carboxylic acid
L07133.14
L07133.22
L07133.14EA
59
EUR
L07133.14
L07133.22
Cumarin-3-carbonsäure ≥98%
Cumarin-3-carbonsäure
Purity: 98%
CAS No.: 531-81-7
Molecular Formula: C10H6O4
Formula Weight: 190.15
Melting Point: 190-193°
Beilstein Reference: 154276
Merck Reference: 14,2563
UN No.: 2811
MDL No.: MFCD00006852
Formel:
C₁₀H₆O₄ Molecular Weight: 190.16 g/mol Schmelzpunkt: 190…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006852 CAS-Nummer: 531-81-7 EINECS: 208-518-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...