2,6-Dimethoxy-p-benzochinon ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Dimethoxy-1,4-Benzoquinone

A14936.03 A14936.06 A14936.14
A14936.03EA 25.9 EUR
A14936.03 A14936.06 A14936.14
2,6-Dimethoxy-p-benzochinon ≥98%
2,6-Dimethoxy-p-benzochinon

Purity: 98% CAS No.: 530-55-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 252-257° Beilstein Reference: 2048182 Merck Reference: 14,3225 MDL No.: MFCD00016368

Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
Schmelzpunkt: 252…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016368
CAS-Nummer: 530-55-2
EINECS: 208-484-7
Merck Index: 14,3225

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR