2,5-Di-tert-butyl-p-benzochinon ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,5-Di-tert-butylcyclohexa-2,5-diene-1,4-dione
L10054.06
L10054.14
L10054.06EA
38.3
EUR
L10054.06
L10054.14
2,5-Di-tert-butyl-p-benzochinon ≥99%
2,5-Di-tert-butyl-p-benzochinon
Purity: 99%
CAS No.: 2460-77-7
Molecular Formula: C14H20O2
Formula Weight: 220.31
Melting Point: 152-153°
Beilstein Reference: 2047945
Formel:
C₁₄H₂₀O₂ Molecular Weight: 220,31 g/mol Schmelzpunkt: 152…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019442 CAS-Nummer: 2460-77-7 EINECS: 219-552-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...