2,5-Dimethyl-p-benzochinon 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,5-Dimethylcyclohexa-2,5-diene-1,4-dione
408230050
408230250
ACRO408230050EA
78
EUR
ACRO408230050
ACRO408230250
2,5-Dimethyl-p-benzochinon 99%
2,5-Dimethyl-p-benzochinon
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041737 CAS-Nummer: 137-18-8 EINECS: 205-283-6 UN: 2811 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 123 to 126 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...