2,5-Dimethyl-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dimethylcyclohexa-2,5-diene-1,4-dione

39579-25G 39579-5G
SIAL39579-25GEA 626 EUR
SIAL39579-25G SIAL39579-5G
2,5-Dimethyl-p-benzochinon, Sigma-Aldrich®
2,5-Dimethyl-p-benzochinon
Formel: C₈H₈O₂
Molecular Weight: 136,15 g/mol
MDL: MFCD00041737
CAS-Nummer: 137-18-8
EINECS: 205-283-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR