2,6-Di-tert-butyl-p-benzochinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,6-Di-tert-butylcyclohexa-2,5-diene-1,4-dione
M-Clarity™ Qualitätsstufe = MQ100
153931-1G
153931-5G
SIAL153931-1GEA
46.7
EUR
SIAL153931-1G
SIAL153931-5G
2,6-Di-tert-butyl-p-benzochinon, Sigma-Aldrich®
2,6-Di-tert-butyl-p-benzochinon
Formel:
C₁₄H₂₀O₂ Molecular Weight: 220,31 g/mol |
MDL:
MFCD00001601 CAS-Nummer: 719-22-2 EINECS: 211-946-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...