2,6-Dimethyl-p-benzochinon ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,6-Dimethylcyclohexa-2,5-diene-1,4-dione
A11342.03
A11342.06
A11342.14
AB116176.0025
A11342.03EA
36.9
EUR
A11342.03
A11342.06
A11342.14
ABCRAB116176.0025
2,6-Dimethyl-p-benzochinon ≥99%
2,6-Dimethyl-p-benzochinon
Purity: 99%
CAS No.: 527-61-7
Molecular Formula: C8H8O2
Formula Weight: 136.15
Melting Point: 69-71°
Beilstein Reference: 2041345
Formel:
C₈H₈O₂ Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001605 CAS-Nummer: 527-61-7 EINECS: 208-420-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...