3,5-Di-tert-butyl-o-benzochinon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L00169.06 L00169.14
L00169.06EA 68.3 EUR
L00169.06 L00169.14
3,5-Di-tert-butyl-o-benzochinon ≥98%
3,5-Di-tert-butyl-o-benzochinon

Purity: 98+% CAS No.: 3383-21-9 Molecular Formula: C14H20O2 Formula Weight: 220.31 Melting Point: 113-117° Beilstein Reference: 2047944

Formel: C₁₄H₂₀O₂
Molecular Weight: 220.31 g/mol
Schmelzpunkt: 113…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001647
CAS-Nummer: 3383-21-9
EINECS: 222-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR