3-Amino-4-brom-1H-pyrazol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

A11012.06 A11012.14
A11012.06EA 187 EUR
A11012.06 A11012.14
3-Amino-4-brom-1H-pyrazol 97%
3-Amino-4-bromo-1H-pyrazole

Purity: 97% CAS No.: 16461-94-2 Molecular Formula: C3H4BrN3 Formula Weight: 161.99 Odour: Odourless Melting Point: 134-137° Beilstein Reference: 1617109 MDL No.: MFCD00082728

Formel: C₃H₄BrN₃
Molecular Weight: 161,99 g/mol
Schmelzpunkt: 134…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082728
CAS-Nummer: 16461-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR