3-Amino-4-brom-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

005504-1G 005504-5G
FLUO005504-1GEA 45.9 EUR
FLUO005504-1G FLUO005504-5G
3-Amino-4-brom-1H-pyrazol
3-Amino-4-bromo-1H-pyrazole
Formel: C₃H₄BrN₃
Molecular Weight: 161.99 g/mol
MDL: MFCD00082728
CAS-Nummer: 16461-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR