1-Methyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

010814-5G 010814-1G
FLUO010814-5GEA 0 EUR
FLUO010814-5G FLUO010814-1G
1-Methyl-1H-pyrazol-4-boronsäurepinakolester
1-Methyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₀H₁₇BN₂O₂
Molecular Weight: 208.07 g/mol
MDL: MFCD03789259
CAS-Nummer: 761446-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR