1-Methyl-1H-pyrazol-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4,4,5,5-Tetramethyl-2-(1-methylpyrazol-5-yl)-1,3,2-dioxaborolane, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methylpyrazole-5-boronic acid Pinacol ester

PN-8865-5G PN-8865-1G
COBBPN-8865-5GEA 51.3 EUR
COBBPN-8865-5G COBBPN-8865-1G
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₀H₁₇BN₂O₂
Molecular Weight: 208.07 g/mol
MDL: MFCD05861380
CAS-Nummer: 847818-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR