3-Amino-5-methyl-1H-pyrazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine
368030250
368031000
368030050
ACRO368030250EA
68.6
EUR
ACRO368030250
ACRO368031000
ACRO368030050
3-Amino-5-methyl-1H-pyrazol 97%
3-Amino-5-methyl-1H-pyrazole
Formel:
C₄H₇N₃ Molecular Weight: 97,12 g/mol Schmelzpunkt: 41…43 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075180 CAS-Nummer: 31230-17-8 |
Spezifikation Testergebnisse
Appearance | Light yellow to orange Solid or viscous liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...