3-Amino-5-methyl-1H-pyrazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine
A11642.06
A11642.14
A11642.22
A11642.06EA
26.3
EUR
A11642.06
A11642.14
A11642.22
3-Amino-5-methyl-1H-pyrazol 97%
3-Amino-5-methyl-1H-pyrazole
Purity: 97%
CAS No.: 31230-17-8
Molecular Formula: C4H7N3
Formula Weight: 97.12
Flash Point:
Formel:
C₄H₇N₃ Molecular Weight: 97,12 g/mol Siedepunkt: 212…213 °C (14 mmHg) Schmelzpunkt: 42…47 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075180 CAS-Nummer: 31230-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...