3-Amino-5-methyl-1H-pyrazol
Lieferant: Thermo Scientific
Synonyme:
3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine
KM00084DA
KM00084EA
MAYBKM00084DAEA
0
EUR
MAYBKM00084DA
MAYBKM00084EA
3-Amino-5-methyl-1H-pyrazol
3-Amino-5-methyl-1H-pyrazole
Formel:
C₄H₇N₃ Molecular Weight: 97,12 g/mol |
MDL:
MFCD00075180 CAS-Nummer: 31230-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...