5-Amino-1-methyl-1H-pyrazol

Lieferant: BIONET RESEARCH

Synonyme: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

4T-0063.500 4T-0063.5 4T-0063.1 4T-0063.10
BIOT4T-0063.500EA 0 EUR
BIOT4T-0063.500 BIOT4T-0063.5 BIOT4T-0063.1 BIOT4T-0063.10
5-Amino-1-methyl-1H-pyrazol
5-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97,12 g/mol
MDL: MFCD00068156
CAS-Nummer: 1192-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR