1-Methyl-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid
BB-8567-5G
BB-8567-1G
COBBBB-8567-5GEA
571
EUR
COBBBB-8567-5G
COBBBB-8567-1G
1-Methyl-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
1-Methyl-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
Formel:
C₅H₆BF₃N₂O₂ Molecular Weight: 193,92 g/mol |
MDL:
MFCD08701785 CAS-Nummer: 344591-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...