3,5-Dimethyl-4-iod-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dimethyl-4-iodopyrazole

M-Clarity™ Qualitätsstufe = MQ100

683523-5G
SIAL683523-5GEA 94.2 EUR
SIAL683523-5G
3,5-Dimethyl-4-iod-1H-pyrazol, Sigma-Aldrich®
3,5-Dimethyl-4-iodo-1H-pyrazole
Formel: C₅H₇IN₂
Molecular Weight: 222.03 g/mol
MDL: MFCD00040247
CAS-Nummer: 2033-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR