5-Brom-2-methoxypyrimidin

Lieferant: COMBI-BLOCKS

PY-7826-25G PY-7826-5G
COBBPY-7826-25GEA 166 EUR
COBBPY-7826-25G COBBPY-7826-5G
5-Brom-2-methoxypyrimidin
5-Bromo-2-methoxypyrimidine
Formel: C₅H₅BrN₂O
Molecular Weight: 189,01 g/mol
MDL: MFCD04973351
CAS-Nummer: 14001-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR