5-Brom-2-hydroxypyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromopyrimidin-2(1H)-one, 5-Bromo-2-pyrimidinone

OS-7586-25G OS-7586-5G
COBBOS-7586-25GEA 127 EUR
COBBOS-7586-25G COBBOS-7586-5G
5-Brom-2-hydroxypyrimidin
5-Bromo-2-hydroxypyrimidine
Formel: C₄H₃BrN₂O
Molecular Weight: 174,98 g/mol
MDL: MFCD01318956
CAS-Nummer: 38353-06-9
EINECS: 253-896-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR