2-Amino-4-chlor-6-methylpyrimidin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Chloro-6-methylpyrimidin-2-amine
L06896.06
L06896.14
L06896.06EA
25
EUR
L06896.06
L06896.14
2-Amino-4-chlor-6-methylpyrimidin 98%
2-Amino-4-chloro-6-methylpyrimidine
Purity: 98%
CAS No.: 5600-21-5
Molecular Formula: C5H6ClN3
Formula Weight: 143.58
Melting Point: 181-186°
Beilstein Reference: 114297
Formel:
C₅H₆ClN₃ Molecular Weight: 143,58 g/mol Schmelzpunkt: 181…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006091 CAS-Nummer: 5600-21-5 EINECS: 227-018-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...