2-Amino-4-hydroxy-6-methylpyrimidin

Lieferant: CHEMPUR

Synonyme: 2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol

KM02128.250 KM02128.1
CHMPKM02128.250EA 0 EUR
CHMPKM02128.250 MAYBKM02128.1
2-Amino-4-hydroxy-6-methylpyrimidin
2-Amino-4-hydroxy-6-methylpyrimidine
Formel: C₅H₇N₃O
Molecular Weight: 125.13 g/mol
MDL: MFCD00006095
CAS-Nummer: 3977-29-5
EINECS: 223-612-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR