Fenclorim

Lieferant: BIONET RESEARCH

Synonyme: 4,6-Dichloro-2-phenylpyrimidine

1J-330S.500 1J-330S.1 1J-330S.5 1J-330S.10
BIOT1J-330S.500EA 0 EUR
BIOT1J-330S.500 BIOT1J-330S.1 BIOT1J-330S.5 BIOT1J-330S.10
Fenclorim
Fenclorim
Formel: C₁₀H₆Cl₂N₂
MDL: MFCD00754193
CAS-Nummer: 3740-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR