Methylphenylsulfoxid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (Methylsulphinyl)benzene

A15009.06 A15009.14 A15009.22
A15009.06EA 44.4 EUR
A15009.06 A15009.14 A15009.22
Methylphenylsulfoxid ≥98%
Methylphenylsulfoxid

Purity: 98+% CAS No.: 1193-82-4 Molecular Formula: C7H8OS Formula Weight: 140.20 Storage Temperature: 2°C to 8°C Flash Point: 86°(186°F) Refractive Index: 1.5775 Melting Point: 26-32° Boiling Point: 93-94°/0.3mm Beilstein Reference: 1904976 MDL No.: MFCD00002088

Formel: C₇H₈OS
Molecular Weight: 140,21 g/mol
Siedepunkt: 93…94 °C (0,3 mmHg)
Schmelzpunkt: 26…32 °C
Flammpunkt: 86 °C (186 °F)
MDL: MFCD00002088
CAS-Nummer: 1193-82-4
EINECS: 214-781-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR