Methylphenylsulfoxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Methylsulphinyl)benzene

M-Clarity™ Qualitätsstufe = MQ200

261696-25G 261696-5G
SIAL261696-25GEA 308 EUR
SIAL261696-25G SIAL261696-5G
Methylphenylsulfoxid, Sigma-Aldrich®
Methylphenylsulfoxid
Formel: C₇H₈OS
Molecular Weight: 140,21 g/mol
MDL: MFCD00002088
CAS-Nummer: 1193-82-4
EINECS: 214-781-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR