2-Amino-5,6-dimethyl-4-hydroxypyrimidin

Lieferant: Thermo Scientific

Synonyme: 2-Amino-5,6-dimethyl-4-pyrimidinol

KM09425.1 KM09425.10
MAYBKM09425.1EA 0 EUR
MAYBKM09425.1 MAYBKM09425.10
2-Amino-5,6-dimethyl-4-hydroxypyrimidin
2-Amino-5,6-dimethyl-4-hydroxypyrimidin
Formel: C₆H₉N₃O
Molecular Weight: 139,16 g/mol
MDL: MFCD00023258
CAS-Nummer: 3977-23-9
EINECS: 223-611-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR