Cholesterylchlorformiat
Lieferant: Thermo Scientific
|
Gefahr
|
L04801.14
L04801.22
L04801.14EA
46.6
EUR
L04801.14
L04801.22
Cholesterylchlorformiat
Cholesterylchlorformiat
CAS No.: 7144-08-3
Molecular Formula: C28H45ClO2
Formula Weight: 449.12
Storage Temperature: 2°C to 8°C
Odour: Pungent
Melting Point: 116-120°
Beilstein Reference: 3223154
Optical Rotation: -29° (c=5 in chloroform)
UN No.: 3261
MDL No.: MFCD00003633
Formel:
C₂₈H₄₅ClO₂ Molecular Weight: 449,12 g/mol Schmelzpunkt: 116…120 °C |
MDL:
MFCD00003633 CAS-Nummer: 7144-08-3 EINECS: 230-447-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...