Cholesterylchlorformiat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
C77007-100G
C77007-25G
SIALC77007-100GEA
225
EUR
SIALC77007-100G
SIALC77007-25G
Cholesterylchlorformiat, Sigma-Aldrich®
Cholesterylchlorformiat
Formel:
C₂₈H₄₅ClO₂ Molecular Weight: 449,12 g/mol |
MDL:
MFCD00003633 CAS-Nummer: 7144-08-3 EINECS: 230-447-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...