2-Methyl-1,3-dioxolan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

292206-25G 292206-5G
SIAL292206-25GEA 76.8 EUR
SIAL292206-25G SIAL292206-5G
2-Methyl-1,3-dioxolan, Sigma-Aldrich®
2-Methyl-1,3-dioxolan
Formel: C₄H₈O₂
Molecular Weight: 88,11 g/mol
MDL: MFCD00003210
CAS-Nummer: 497-26-7
EINECS: 207-841-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR