(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
A18499.22
A18499.36
A18499.22EA
54
EUR
A18499.22
A18499.36
(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid ≥98%
D-Panthenol
Purity: 98+%
CAS No.: 81-13-0
Molecular Formula: C9H19NO4
Formula Weight: 205.26
Storage Temperature: 2°C to 8°C
Flash Point: 159°(318°F)
Refractive Index: 1.4990
Boiling Point: 118-120°/0.02m
Beilstein Reference: 1724947
Merck Reference: 14,2947
Optical Rotation: -30° (c=5 in water)
MDL No.: MFCD00065006
Formel:
C₉H₁₉NO₄ Molecular Weight: 205.25 g/mol Siedepunkt: 118…120 °C (0.02 mmHg) Dichte: 1.162 Flammpunkt: 159 °C (318 °F) |
MDL:
MFCD00065006 CAS-Nummer: 81-13-0 EINECS: 201-327-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...