α-Ketoglutaric acid disodium salt dihydrate, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Oxoglutaric acid disodium salt dihydrate, α-Ketoglutaric acid disodium salt dihydrate, Sodium α-ketoglutarate dibasic dihydrate
75892-100G
75892-25G
SIAL75892-100GEA
151
EUR
SIAL75892-100G
SIAL75892-25G
α-Ketoglutaric acid disodium salt dihydrate, Sigma-Aldrich®
α-Ketoglutaric acid disodium salt dihydrate
Formel:
C₅H₄O₅·2Na·2H₂O Molecular Weight: 226,092 g/mol |
MDL:
MFCD00150702 CAS-Nummer: 305-72-6 EINECS: 206-167-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...