(Phenylthio)essigsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
B22280.14
B22280.22
B22280.14EA
62.5
EUR
B22280.14
B22280.22
(Phenylthio)essigsäure ≥97%
(Phenylthio)essigsäure
Purity: 97%
CAS No.: 103-04-8
Molecular Formula: C6H5SCH2CO2H
Formula Weight: 168.22
Odour: Stench
Melting Point: 61-66°
Beilstein Reference: 2045072
MDL No.: MFCD00004355
Formel:
C₈H₈O₂S Molecular Weight: 168,22 g/mol Schmelzpunkt: 61…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004355 CAS-Nummer: 103-04-8 EINECS: 203-073-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...