Bismut(III)citrat ≥94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 94%
CAS No.: 813-93-4
Molecular Formula: C6H5BiO7
Formula Weight: 398.08
Physical Form: Crystalline Powder, typically 10 micron max
Density: 3.5
Appearance: White
Odour: Odourless
Melting Point: dec.
Solubility: Insoluble in water, alcohol, ether. Soluble in ammonia solution and in solution of alkali citrates. Typical tap density 0.50g/cm3
MDL No.: MFCD00050817
Formel:
C₆H₅BiO₇ Dichte: 3,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00050817 CAS-Nummer: 813-93-4 EINECS: 212-390-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...