(4-Fluorphenylthio)essigsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-((4-Fluorophenyl)thio)acetic acid

A19658.03 A19658.06
A19658.03EA 41.5 EUR
A19658.03 A19658.06
(4-Fluorphenylthio)essigsäure ≥97%
(4-Fluorphenylthio)essigsäure

Purity: 97% CAS No.: 332-51-4 Molecular Formula: C8H7FO2S Formula Weight: 186.21 Melting Point: 76-79° MDL No.: MFCD00219825

Formel: C₈H₇FO₂S
Molecular Weight: 186,21 g/mol
Schmelzpunkt: 76…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00219825
CAS-Nummer: 332-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR