(4-Fluorphenylthio)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-((4-Fluorophenyl)thio)acetic acid

006343-1G 006343-5G
FLUO006343-1GEA 55.4 EUR
FLUO006343-1G FLUO006343-5G
(4-Fluorphenylthio)essigsäure
(4-Fluorphenylthio)essigsäure
Formel: C₈H₇FO₂S
Molecular Weight: 186,21 g/mol
MDL: MFCD00219825
CAS-Nummer: 332-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR