Bis(phenylthio)methan
Lieferant: FLUOROCHEM
003674-25G
003674-100G
FLUO003674-25GEA
69.8
EUR
FLUO003674-25G
FLUO003674-100G
Bis(phenylthio)methan
Bis(phenylthio)methan
Formel:
C₁₃H₁₂S₂ Molecular Weight: 232,37 g/mol |
MDL:
MFCD00003067 CAS-Nummer: 3561-67-9 EINECS: 222-624-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...