1-Benzyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

PN-8624-1G PN-8624-5G
COBBPN-8624-1GEA 73.6 EUR
COBBPN-8624-1G COBBPN-8624-5G
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₆H₂₁BN₂O₂
Molecular Weight: 284.16 g/mol
MDL: MFCD03789252
CAS-Nummer: 761446-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR