1-Benzyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

M-Clarity™ Qualitätsstufe = MQ100

636002-1G 636002-5G
SIAL636002-1GEA 41.6 EUR
SIAL636002-1G SIAL636002-5G
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
1-Benzyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₆H₂₁BN₂O₂
Molecular Weight: 284,16 g/mol
MDL: MFCD03789252
CAS-Nummer: 761446-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR