1-Benzyl-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-Benzyl-1H-pyrazol-4-yl)boronic acid

BB-8452-5G BB-8452-1G
COBBBB-8452-5GEA 1650 EUR
COBBBB-8452-5G COBBBB-8452-1G
1-Benzyl-1H-pyrazol-4-boronsäure
1-Benzyl-1H-pyrazol-4-boronsäure
Formel: C₁₀H₁₁BN₂O₂
Molecular Weight: 202,02 g/mol
MDL: MFCD04038760
CAS-Nummer: 852362-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR