(3-Acryloxypropyl)trimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Trimethoxysilyl)propyl acrylate

SILS00155.5 SILS00155.25
SILNSILS00155.5EA 0 EUR
SILNSILS00155.5 SILNSILS00155.25
(3-Acryloxypropyl)trimethoxysilan
(3-Acryloxypropyl)trimethoxysilan
Formel: C₉H₁₈O₅Si
Molecular Weight: 234.32 g/mol
MDL: MFCD00054803
CAS-Nummer: 4369-14-6
EINECS: 419-560-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR