4-(4-Bromphenoxy)phenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

695351-25G 695351-5G
SIAL695351-25GEA 428 EUR
SIAL695351-25G SIAL695351-5G
4-(4-Bromphenoxy)phenol, Sigma-Aldrich®
4-(4-Bromphenoxy)phenol
Formel: C₁₂H₉BrO₂
Molecular Weight: 265,11 g/mol
MDL: MFCD08446910
CAS-Nummer: 13320-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR