Succinylsulfathiazol
Lieferant: MP Biomedicals
Synonyme:
N4-Succinyl-N-(2-thiazolyl)sulphanilamide, 4-(Succinylamino)-N-(2-thiazolyl)benzenesulphonamide, 4-Oxo-4-((4-(N-(thiazol-2-yl)sulphamoyl)phenyl)amino)butanoic acid
0210297680
0210297690
ICNA0210297680EA
110
EUR
ICNA0210297680
ICNA0210297690
Succinylsulfathiazol
Succinylsulfathiazol
Formel:
C₁₃H₁₃N₃O₅S₂ Molecular Weight: 355,38 g/mol |
MDL:
MFCD00022437 CAS-Nummer: 116-43-8 EINECS: 204-141-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...